Learn More
DWK Life Sciences Wheaton™ Glass Sample Vials in Lab File without Caps
Packaged in partitioned trays, without caps
Supplier:  DWK Life Sciences 224832
 View more versions of this product
Catalog No. 06451173
Only null left
Description
Description
Partitioned trays provide an easy way to inventory samples or to store empty vials.
  • Autoclavable
  • Clear or Amber Vials
    • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
    • Amber vials made from borosilicate glass that protects light-sensitive products
    Order Info
    Caps available separately
    Compliance
    Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements. Amber vials conform to USP Type 1 requirements for light transmission.
    Specifications
    Chemical Identifiers
    Borosilicate Glass
    Screw Cap
    Autoclavable
    Screw
    M-T File™ and Vial Rack
    Specifications
    Clear
    Borosilicate Glass
    Partitioned Tray
    6.76 dr.
    24-400
    Autoclavable
    70 mm
    28 mm
    25 mL
    Screw
    Screw Cap
    M-T File™ and Vial Rack
    ASTM E438, USP Type I, Type I, Class A
    Sample Vial in Lab File
    Product Suggestions
    Product Suggestions