Learn More
DWK Life Sciences Wheaton™ E-Z Decappers
With ergonomically designed handles for sealing and decapping standard laboratory vials
$938.44 - $1,106.90
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|
06451461
|
DWK Life Sciences
W225351 |
11 mm Seals |
Each for $1,099.28
|
|
|||||
|
06451462
|
DWK Life Sciences
W225352 |
13 mm Seals |
Each for $1,106.90
|
|
|||||
|
06451463
|
DWK Life Sciences
W225353 |
20 mm Seals |
Each for $938.44
|
|
|||||
Description
Wheaton E-Z Decappers are recommended for decapping standard laboratory vials.
- Several sizes of decapper are available, including those that remove seals 8mm, 11mm, 13mm and 20mm wide.
- The decappers come with visible size labels for easy identification.
- User comfort is preserved by an ergonomically designed, cushioned handle. Hand fatigue is reduced by the short handle and redesigned spring assembly.
- Decappers can be autoclaved, although autoclaving may change the appearance or reduce product life. They should not be autoclaved at greater than 121°C
Warranty
Two years