Learn More
DWK Life Sciences DURAN™ PURE Clear Glass Bottles
Primary packaging products for specific requirements in pharmaceutical industries.
$384.27 - $4,435.21
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
13710912
|
DWK Life Sciences
818011401 |
25 mL |
Case of 10 Each for $384.27
|
|
|||||
|
13710913
|
DWK Life Sciences
818012406 |
100 mL |
Case of 10 Each for $464.30
|
|
|||||
|
13710914
|
DWK Life Sciences
818013608 |
250 mL |
Case of 10 Each for $507.71
|
|
|||||
|
13710915
|
DWK Life Sciences
818014407 |
500 mL |
Case of 10 Each for $563.56
|
|
|||||
|
13710916
|
DWK Life Sciences
818015403 |
1000 mL |
Case of 10 Each for $800.76
|
|
|||||
|
13710917
|
DWK Life Sciences
818016305 |
2000 mL |
Case of 10 Each for $1,669.68
|
|
|||||
|
13710918
|
DWK Life Sciences
818017301 |
5000 mL |
Each for $1,009.29
|
|
|||||
|
13710919
|
DWK Life Sciences
818018606 |
10 L |
Each for $1,940.44
|
|
|||||
|
13710920
|
DWK Life Sciences
818019105 |
20 L |
Each for $4,435.21
|
|
|||||
Description
- Made of borosilicate 3.3 neutral Glass Type 1
- Includes protective dust covers
- Graduated
- Do not include screw caps
- Available in volumes ranging from 25mL to 20L