missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Disposable Drum Thieves

Economical and convenient to use

Supplier:  DWK Life Sciences 885300

 View more versions of this product

Catalog No. 1420972

Only null left

Description

Description

  • Simply insert empty tube into drum, hold thumb over the top, and withdraw sample
  • Soda-lime glass units break easily for quick disposal
  • Length: 43 in. (109cm)

Specifications

Chemical Identifiers

Drum Thief
Single-use
43 in.
Soda Lime Glass
Ideal for quick and easy sample retrieval
109.2 cm

Specifications

Drum Thief
25 mL
Ideal for quick and easy sample retrieval
0.75 cm
109.2 cm
Soda Lime Glass
Single-use
0.29 in.
43 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions