Learn More
DWK Life Sciences™ Wheaton™ Caps for 12 x 32 mm Large Opening ABC Vials
These closures for ABC Vials feature the Wheaton™ Super Septa, which is low extractable and optimal for GC/MS.
Supplier: DWK Life Sciences™ W2253320207
Description
- Screw caps are available in natural, black, red, blue, yellow or green polypropylene
Chemical Identifiers
| Polypropylene | |
| 100 Caps/Pk., 10 Pk./Cs. |
Specifications
| Green | |
| No | |
| 100 Caps/Pk., 10 Pk./Cs. |
| Polypropylene | |
| PTFE, Silicone |