missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ BOD Bottles

Catalog No. 0292633
Change view
Click to view available options
Volume (Metric):
60 mL
300 mL
No. Sequence:
1 to 24
1 to 36
121 to 144
145 to 168
169 to 192
193 to 216
217 to 240
241 to 264
25 to 48
265 to 288
289 to 312
313 to 336
337 to 360
361 to 384
385 to 408
409 to 432
49 to 72
73 to 96
97 to 120
Specials
Unnumbered
Stopper:
Pennyhead
Robotic
Barcode:
No
Yes
35 product options available for selection
Product selection table with 35 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) No. Sequence Stopper Barcode
0292633 300 mL 145 to 168 Robotic Yes
02912547 60 mL Specials Robotic Yes
02912549 300 mL 289 to 312 Pennyhead Yes
0292611 300 mL 49 to 72 Pennyhead Yes
0292612 60 mL Unnumbered Robotic No
0292613 60 mL 1 to 36 Robotic Yes
0292625 300 mL 73 to 96 Pennyhead Yes
0292626 300 mL Unnumbered Robotic Yes
0292627 300 mL 1 to 24 Robotic Yes
0292628 300 mL 25 to 48 Robotic Yes
0292629 300 mL 49 to 72 Robotic Yes
0292630 300 mL 73 to 96 Robotic Yes
0292632 300 mL 121 to 144 Robotic Yes
0292631 300 mL 97 to 120 Robotic Yes
0292634 300 mL 169 to 192 Robotic Yes
0292635 300 mL 193 to 216 Robotic Yes
0292636 300 mL 217 to 240 Robotic Yes
0292637 300 mL 241 to 264 Robotic Yes
0292639 300 mL 289 to 312 Robotic Yes
0292638 300 mL 265 to 288 Robotic Yes
0292640 300 mL 313 to 336 Robotic Yes
0292641 300 mL 337 to 360 Robotic Yes
0292642 300 mL 361 to 384 Robotic Yes
0292643 300 mL 385 to 408 Robotic Yes
0292644 300 mL 409 to 432 Robotic Yes
029268 300 mL 1 to 24 Pennyhead Yes
029267A 300 mL Unnumbered Pennyhead Yes
029269 300 mL 25 to 48 Pennyhead Yes
0292690 300 mL 97 to 120 Pennyhead Yes
0292691 300 mL 121 to 144 Pennyhead Yes
0292692 300 mL 145 to 168 Pennyhead Yes
0292693 300 mL 169 to 192 Pennyhead Yes
0292694 300 mL 193 to 216 Pennyhead Yes
0292695 300 mL 217 to 240 Pennyhead Yes
0292696 300 mL 241 to 264 Pennyhead Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 35 options available.
35 options
Catalog No. 0292633 Supplier DWK Life Sciences Supplier No. 22749707

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Available with either glass robotic or pennyhead stoppers

Bottles

  • Borosilicate glass, 60 or 300mL, numbered or unnumbered
  • With permanent marking areas; numbered versions are also bar-coded
  • Specially designed shoulder radius sweeps all air from inside the bottle during filling
  • 300mL bottles are recommended for the five-day BOD test referenced in Standard Methods for the Examination of Water and Wastewater, 18th Edition, 5210B, 1992
Stoppers
  • Interchangeable borosilicate glass stoppers have tapered bottoms that also prevent air entrapment
  • Flared mouth of bottle forms a water seal around stoppers that prevents air from being drawn in during incubation
  • Stopper joint is compatible with the probes of the leading BOD and DO meters
  • Glass robotic stoppers are compatible with most robotic systems

Use to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action.

Order Info

Dimensions: 300mL bottles are 2.75 diameter x 6.5 in. high (69 x 165mm); 60mL bottles are 1.75 diameter x 4.5 in. high (43 x 115mm)

Specifications

Barcode Yes
Volume (Metric) 300 mL
For Use With (Application) Used to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action
Marking Spot Yes
Material Borosilicate Glass
No. Sequence 145 to 168
Stopper Robotic
Volume (English) 10.14 oz.
Height (English) 6.49 in.
Height (Metric) 16.5 cm
Diameter (English) 2.7 in.
Diameter (Metric) 6.9 cm
Show More Show Less