Learn More
DWK Life Sciences Wheaton™ Black Phenolic Screw Cap with PTFE-Faced Rubber Liner
Features PTFE faced (14B) styrene-butadiene rubber liner
Supplier:  DWK Life Sciences 240421
 View more versions of this product
Catalog No. 0645050
Only null left
Description
Description
  • Designed for product safety
  • TFE/Styrene-Butadiene rubber liner
  • Screw cap
  • Pheolic
  • Black
  • Autoclavable
Specifications
Chemical Identifiers
Black
ISO 9001
PTFE/14B Rubber
Phenolic Resin
Bottle
Autoclavable
Specifications
Black
100 Cs.
ISO 9001
PTFE/14B Rubber
Phenolic Resin
33-400
Bottle
Autoclavable
Product Suggestions
Product Suggestions