missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Black Phenolic Open Top Cap with Gray Butyl Septa and Flange
Replacement Black Phenolic Screw Caps for Media Bottles
$338.22 - $5,364.86
Specifications
| Color | Black |
|---|---|
| Includes | Gray Butyl septa and flange |
| Material | Phenolic Resin |
| Neck Finish | 33-430 |
| Certifications/Compliance | ISO 9001 |
Description
Replacement Black Phenolic Screw Caps for Media Bottles
Ideal for use with Wheaton media bottles
- Open top screw caps
- Size: 33–430
- Gray Chlorobutyl/50 septa
- Phenolic
- Black
- Autoclavable
Specifications
| Black | |
| Phenolic Resin | |
| ISO 9001 | |
| Bottle |
| Gray Butyl septa and flange | |
| 33-430 | |
| Open Top Cap |