missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Open Top Cap with Gray Butyl Septa and Flange

Catalog No. 0645075
Change view
Click to view available options
No. per Case:
100 Case
1000 Case
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Case
0645075 1000 Case
0645074 100 Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0645075 Supplier DWK Life Sciences Supplier No. 240683

Replacement Black Phenolic Screw Caps for Media Bottles

  • Open Top Cap With Gray Chlorobutyl / 50 Septa and Flange
  • Autoclavable

Specifications

Color Black
Includes Gray Butyl septa and flange
Material Phenolic Resin
No. per Case 1000 Case
Neck Finish 33-430
Certifications/Compliance ISO 9001
Product Type Open Top Cap
For Use With (Equipment) Bottle
Autoclavable Autoclavable