missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Cap with Rubber Liner

14B rubber-lined caps feature a nontoxic, white, styrene-butadiene rubber liner, ideal for cell culture work

$278.89 - $468.96

Specifications

Color Black
Material Phenolic Resin
No. per Case 1000 Cs.
Certifications/Compliance ISO 9001
For Use With (Equipment) Bottle
View More Specs

Products 4
Catalog Number Mfr. No. Neck Finish Price Quantity  
Catalog Number Mfr. No. Neck Finish Price Quantity  
02-912-249
View Documents
DWK Life Sciences
W239302
33-400
Case of 144 Each for $361.86
Only null left
 
02-912-251
View Documents
DWK Life Sciences
W239304
43-400
Case of 72 Each for $278.89
Only null left
 
02-912-252
View Documents
DWK Life Sciences
W239306
48-400
Case of 72 Each for $468.96
Only null left
 
03-410-302
View Documents
DWK Life Sciences
239301
28-400
Case of 144 Each for $375.57
Only null left
 
Description

Description

Specifications

Specifications

Black
1000 Cs.
Bottle
Phenolic Resin
ISO 9001
Rubber
Videos
SDS
Documents

Documents