missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Cap with Rubber Liner

Catalog No. 03410302
Change view
Click to view available options
Neck Finish:
28-400
33-400
43-400
48-400
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish
03410302 28-400
02912249 33-400
02912251 43-400
02912252 48-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03410302 Supplier DWK Life Sciences Supplier No. 239301

14B rubber-lined caps feature a nontoxic, white, styrene-butadiene rubber liner, ideal for cell culture work

Specifications

Color Black
Material Phenolic Resin
No. per Case 1000 Cs.
Neck Finish 28-400
Certifications/Compliance ISO 9001
Closure Included Yes
Product Type Phenolic Cap with Rubber Liner
For Use With (Equipment) Bottle
Closure Liner Rubber
Closure Material Phenolic