missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Amber Serum Vials, 10 mL, Sold by MilliporeSigma™ Supelco™

Catalog No. p-7213849
Change view
Click to view available options
No. per Pack:
288/Pk.
36/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
111011397 36/Pk.
111009664 288/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111011397 Supplier Merck Emd Millipore Supplier No. 33143
Only null left

Serum Bottles have a 20 mm, crimp top opening. Closures are not included.

Specifications

Description No Closure Included
Diameter (Metric) Opening 20 mm
Color Amber
Diameter (Metric) Outer 25 mm
Material Glass
Volume (Metric) 10 mL
No. per Pack 36/Pk.
Closure Type Crimp Cap
Autoclavable Autoclavable
Height (Metric) 50 mm
Product Type Serum Bottle
Show More Show Less

WHEATON is a registered trademark of Wheaton Industries, Inc.