missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ Amber Glass Reagent Bottle with Ground Glass Stopper

Catalog No. p-45310
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02911306 100 mL
22308622 250 mL
02918B 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02911306 Supplier DWK Life Sciences Supplier No. W215255

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

WHEATON™ Amber Glass Reagent Bottle with Ground Glass Stopper is designed for safe shelf storage of light-sensitive reagents. Made from amber borosilicate glass with a narrow mouth and a ground-glass octagon flat stopper for a secure, dust-resistant seal.

  • Amber borosilicate glass bottle with narrow mouth
  • Protects light-sensitive reagents and solutions from degradation
  • Ground-glass octagon flat stopper provides an excellent seal and protects the top lip of the bottle from dust

Specifications

Capacity (Metric) 100 mL
Color Amber
Diameter (English) 2.08 in.
Material Glass
Sterility Non-sterile
Closure Color Amber
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 115 mm
Height (English) 4.52 in.
Closure Material Borosilicate Glass
Diameter (Metric) 53 mm
Autoclavable Autoclavable
Capacity (English) 3.38 oz.
Shape Round
Disposable Reusable
Product Type Narrow Mouth Stoppered Bottle
Show More Show Less