Learn More
DWK Life Sciences Acura™ Manual Micropipet Trio Pack
Ready to use set of three manual adjustable pipets
Supplier:  DWK Life Sciences W810323
 View more versions of this product
Catalog No. 13710904
Only null left
Includes: Three Acura manual adjustable pipettes, tip samples, QC certificate and operating instructions
Description
Description
  • Available in nine different Triopacks of specific volumes between 0.1μL to 10mL
Specifications
Chemical Identifiers
Autoclavable
Acura
Three Acura™ manual adjustable pipette, tip samples, QC certificate and operating instructions
825/835
Micropipette Triopack
3 Years
Specifications
Autoclavable
Acura
1 to 10 μL, 10 to 100 μL, 100 to 1000 μL
3 Years
825/835
Micropipette Triopack
Three Acura™ manual adjustable pipette, tip samples, QC certificate and operating instructions
Product Suggestions
Product Suggestions