missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Acura™ Manual Micropipet Trio Pack

Catalog No. 13710901
Change view
Click to view available options
Volume (Metric):
0.1 to 2 μL, 0.5 to 10 μL, 5 to 50 μL
0.1 to 2 μL, 2 to 20 μL, 20 to 200 μL
0.5 to 10 μL, 10 to 100 μL, 100 to 1000 μL
1 to 10 μL, 10 to 100 μL, 100 to 1000 μL
1 to 10 μL, 5 to 50 μL, 20 to 200 μL
10 to 100 μL, 100 to 1000 μL, 0.1 to 5 mL
100 to 1000 μL, 0.5 to 5 mL, 1 to 10 mL
2 to 20 μL, 20 to 200 μL, 100 to 1000 μL
5 to 50 μL, 20 to 200 μL, 100 to 1000 μL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13710901 0.1 to 2 μL, 2 to 20 μL, 20 to 200 μL
13710900 0.1 to 2 μL, 0.5 to 10 μL, 5 to 50 μL
13710902 1 to 10 μL, 5 to 50 μL, 20 to 200 μL
13710903 0.5 to 10 μL, 10 to 100 μL, 100 to 1000 μL
13710905 2 to 20 μL, 20 to 200 μL, 100 to 1000 μL
13710904 1 to 10 μL, 10 to 100 μL, 100 to 1000 μL
13710908 100 to 1000 μL, 0.5 to 5 mL, 1 to 10 mL
13710906 5 to 50 μL, 20 to 200 μL, 100 to 1000 μL
13710907 10 to 100 μL, 100 to 1000 μL, 0.1 to 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 13710901 Supplier DWK Life Sciences Supplier No. W810320

Includes

Three Acura manual adjustable pipettes, tip samples, QC certificate and operating instructions

Ready to use set of three manual adjustable pipets

  • Available in nine different Triopacks of specific volumes between 0.1μL to 10mL

Specifications

Autoclavable Autoclavable
Model 825/835
Product Line Acura
Type Micropipette Triopack
Volume (Metric) 0.1 to 2 μL, 2 to 20 μL, 20 to 200 μL
For Use With (Application) Designed for the accurate, precise pipetteting of solutions, reagents, acids, serums and other liquids and for gel-sequencing applications
Includes Three Acura™ manual adjustable pipette, tip samples, QC certificate and operating instructions
Warranty 3 Years