Learn More
DWK Life Sciences Acura™ electro 956 Eight-Channel Pipets
Fits any single channel volumetric module
$4,258.09 - $5,475.91
Specifications
Battery Type Rechargeable
Compatible Tips Universal fit pipette tip
Model 956
Number of Channels 8
Product Line Acura Electro
View More Specs
Includes: Microprocessor-controlled motor, rechargeable battery pack, QC certificate, operating instructions and samples of pipet tips
Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
03410113
View Documents Promotion Details
DWK Life Sciences
W870926
2.5 to 50 μL
Each for $4,258.09
Only null left
03410115
View Documents Promotion Details
DWK Life Sciences
W870930
20 to 350 μL
Each for $5,475.91
Only null left
03410114
View Documents Promotion Details
DWK Life Sciences
W870928
10 to 200 μL
Each for $5,047.21
Only null left
Description
Description
The microprocessor controlled pipette features intuitive software, eliminating any complicated programming steps. There are six working modes that are easy to access and many volumes to select from.
  • For volume range 0.5 to 350μL
  • Lightweight and perfect hand fitting
  • Sequential tip ejection
Specifications
Specifications
Rechargeable
956
Acura Electro
Digital
Microprocessor controlled motor, rechargeable battery pack, QC certificate, operating instructions and samples of pipette tip
Universal fit pipette tip
8
Pipette
Ergonomic
2 Years