Learn More
DWK Life Sciences Acura™ Electro 926XS Single Channel Micropipet
Catalog No. 03410106
Change view
Click to view available options
Volume (Metric):
0.1 to 2 μL
0.5 to 10 μL
1 to 20 μL
10 to 200 μL
2.5 to 50 μL
5 to 100 μL
50 to 1000 μL
Compatible Tips:
1000μL Pipette tip
200 μL Pipette tip
200μL Pipette tip
Ultra 10 μL Pipette Tip
Ultra 10μL pipette tip
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Compatible Tips
03410106 5 to 100 μL 200μL Pipette tip
03410101 0.1 to 2 μL Ultra 10μL pipette tip
03410103 0.5 to 10 μL 200 μL Pipette tip
03410102 0.5 to 10 μL Ultra 10 μL Pipette Tip
03410104 1 to 20 μL 200μL Pipette tip
03410105 2.5 to 50 μL 200μL Pipette tip
03410107 10 to 200 μL 200μL Pipette tip
03410108 50 to 1000 μL 1000μL Pipette tip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 03410106 Supplier DWK Life Sciences Supplier No. W870912
Only null left
Unsurpassed metrological performance and stability
  • Reduced shaft length and conical end improve driveability
  • Easy access to microtubes and microplates
  • Optional longer volumetric modules
  • Simple conversion by user at any time
  • Optimal ergonomics, light weight
  • Easy, intuitive, self-teaching program
  • Large display, reversible for right or left reading
  • Swift-interchange battery pack
  • Fast charge, long working autonomy
  • Adjustable tip ejector fitting most tips
  • Pipetting cycle counter
  • Speed selector allows swift speed change even during pipetting process
  • Easy tip ejection
Certifications
CE Marked
Compliance
  • Conforms to IVD
  • RoHS compliant
Warranty
Two-years
Specifications
Accuracy ±1 to <±0.5%
Battery Type NiMH
Compatible Tips 200μL Pipette tip
For Use With Pipette Tip 200 μL
Model 926XS
Number of Channels 1
Product Line Acura Electro
Programmability Self-teaching
Type Micropipette
Volume (Metric) 5 to 100 μL
Display Digital
Finger Style Ergonomic
For Use With (Equipment) 200μL Pipette tip
Includes Rechargeable battery pack, individual QC certificate, pipette tip samples and operating instructions
Increments 0.5μL
Warranty 2 Years
Show More Show Less