Learn More
DWK Life Sciences Wheaton™ Accessories and Replacement Parts for 20-Slide Glass Staining Dish
Featuring an open bottom to facilitate rapid immersion and drainage, reducing carryover. Wheaton™ Accessories and Replacement Parts for 20-Slide Glass Staining Dish are manufactured from soda-lime glass.
Supplier:  DWK Life Sciences 900204
 View more versions of this product
Catalog No. 088121B
Only null left
Description
Description
Featuring an open bottom to facilitate rapid immersion and drainage, reducing carryover. Wheaton™ Accessories and Replacement Parts for 20-Slide Glass Staining Dish are manufactured from soda-lime glass.
  • Replacement dish and cover
  • Approx. inside bottom dimensions: 91 x 71 x 60mm
Specifications
Chemical Identifiers
Soda Lime Glass
Specifications
Soda Lime Glass
Slide Staining Rack
Glass Staining Dish
10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back-to-back
Product Suggestions
Product Suggestions