Learn More
DWK Life Sciences Wheaton™ 190mm Soxhlet Extractor With 24/40 Bottom Inner Joint and 34/45 Top Outer Joint
Catalog No. 02912571
Change view
Click to view available options
Certifications/Compliance:
ASTM E 438
Catalog No. Certifications/Compliance
02912571 ASTM E 438
1 options
Catalog No. 02912571 Supplier DWK Life Sciences Supplier No. DWK415000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For continuous extraction of analytes from solid into organic solvent
  • For Soxhlet extraction apparatus
  • Wetted parts manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
  • Size: 30 x 190mm
  • Case of one
Specifications
Product Type Soxhlet Extraction Apparatus
For Use With (Equipment) Soxhlet Extraction Apparatus
For Use With (Application) Used for continuous extraction of analytes from a solid into an organic solvent
Certifications/Compliance ASTM E 438