Learn More
DWK Life Sciences Wheaton™ 13 mm Unlined Aluminum Seal Flip-Cap
Open top seals expose stopper for sample retrieval with a syringe
$45.32
Specifications
| Material (Liner) | Unlined Seal |
|---|---|
| Material | Aluminum |
| Size (Metric) Opening | 13 mm |
| Product Type | Flip Cap |
Description
- Tear-off seal removes completely allowing content to pour from bottle
- Solid top seals for long-term storage of samples
- Flip cap seals provide tamper evidence
Specifications
| Unlined Seal | |
| Aluminum | |
| 13 mm | |
| Flip Cap |