missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Seed Testing Papers: Grade 3014 Sheets
Grade 3014 seed testing pleated strips for testing particularly sensitive seeds.
Supplier: Cytiva 10344672
Description
Grade 3014 seed testing pleated strips for testing particularly sensitive seeds.
Owing to their extremely high purity our seed testing papers provide reliable and reproducible results. The papers are made from pure cellulose without any additives and do not contain any substances that could influence the growth of the seeds. The constant water absorption of the papers ensures the continuous provision of the required amount of water.Specifications
| Sheet | |
| Seed testing | |
| 1000/Pk. | |
| 110 x 20 mm | |
| 20 mm |
| Cellulose | |
| 3014 | |
| Seed Test Paper | |
| 110 mm |