Learn More
Cytiva Whatman™ Reeve Angel 802 Prepleated Qualitative Circles
Coarse retention
Supplier:  Cytiva 5802185
 View more versions of this product
Catalog No. 09901C
Only null left
Description
Description
  • Folded for fast filtration of coarse particles and gelatinous precipitates
  • Qualitative Grade; nominal retention 30-35μm; 0.26mm thick Pleating and creped surface increase effective area, improving both filtering speed and extending filter life
  • Wet strengthened†
Specifications
Chemical Identifiers
Disc
30 to 35 μm
Coarse
15 μm
802
0.26 mm
Specifications
802 18.5 cm 100/pk
15 μm
A prepleated filter for use with a conical filter funnel, offers fast filtration and high loading capacity for analysis involving coarse particles or gelatinous precipitates.
Coarse
0.26 mm
18.5 cm
Disc
30 to 35 μm
802
100/Pk.
18.50 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories