missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Qualitative Filter Paper: Grade 2 Circles

Retains and absorbs more than Grade 1 filters, with a corresponding increase in filtration time. Whatman™ Qualitative Filter Paper: Grade 2 Circles are used for monitoring specific contaminants in the atmosphere and in soil testing.

$25.89 - $304.00

Specifications

Format Disc
Material Cellulose
Particle Retention >8 μm
Filtration Speed 240 sec.
Flow Rate Medium
View More Specs

Products 13
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
09-810A
Accessories View Documents
Cytiva
1002042
42.5 mm
Pack of 100 Each for $35.57
Only null left
 
09-927-156
View Documents
Cytiva
1002047
47 mm
Pack of 100 Each for $25.89
Only null left
 
09-810B
Accessories View Documents
Cytiva
1002055
55 mm
Pack of 100 Each for $28.13
Only null left
 
09-810C
Accessories View Documents
Cytiva
1002070
70 mm
Pack of 100 Each for $30.96
Only null left
 
09-810D Edge
Accessories View Documents
Cytiva
1002090
90 mm
Pack of 100 Each for $43.91
Only null left
Edge
 
09-810E
Accessories View Documents
Cytiva
1002110
110 mm
Pack of 100 Each for $44.48
Only null left
 
09-810F
Accessories View Documents
Cytiva
1002125
125 mm
Pack of 100 Each for $52.99
Only null left
 
09-810G
Accessories View Documents
Cytiva
1002150
150 mm
Pack of 100 Each for $69.18
Only null left
 
09-810H
Accessories View Documents
Cytiva
1002185
185 mm
Pack of 100 Each for $101.00
Only null left
 
09-810J
Accessories View Documents
Cytiva
1002240
240 mm
Pack of 100 Each for $156.45
Only null left
 
09-810K
Accessories View Documents
Cytiva
1002270
270 mm
Pack of 100 Each for $160.89
Only null left
 
09-810L
Accessories View Documents
Cytiva
1002320
320 mm
Pack of 100 Each for $248.04
Only null left
 
09-810M
Accessories View Documents
Cytiva
1002385
385 mm
Pack of 100 Each for $304.00
Only null left
 
Description

Description

Retains and absorbs more than Grade 1 filters, with a corresponding increase in filtration time. Whatman™ Qualitative Filter Paper: Grade 2 Circles are used for monitoring specific contaminants in the atmosphere and in soil testing.

These cellulose filters are used in qualitative analytical techniques to determine and identify materials and for clarifying liquids. 
  • Particle Retention: 8μm
  • Porosity: Medium fine
  • Filtration speed: ASTM™, 55 sec.; Herzberg, 240 sec.
  • Surface: Smooth
  • Holds soil nutrient in plant growth trials
  • Used for soil analysis and seed testing procedures in the agricultural industry
Specifications

Specifications

Disc
>8 μm
Medium
Grade 2
100 Pk.
Cellulose
240 sec.
A standard grade filter used for monitoring specific contaminants in the atmosphere and in soil testing.
Medium Fine
190 μm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'