missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polycap HD Disposable Capsules, 150 HD

Catalog No. 09927245
Change view
Click to view available options
Pore Size:
0.45 μm
5 μm
10 μm
20 μm
Inlet Style:
With 1/2 in. hose barb (HB) inlet/outlet
With Female NPT inlet/outlet
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Inlet Style
09927245 0.45 μm With Female NPT inlet/outlet
09927352 5 μm With Female NPT inlet/outlet
09927684 20 μm With Female NPT inlet/outlet
09927687 10 μm With 1/2 in. hose barb (HB) inlet/outlet
09927685 0.45 μm With 1/2 in. hose barb (HB) inlet/outlet
09927916 10 μm With Female NPT inlet/outlet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 09927245 Supplier Cytiva Supplier No. 2810T
Only null left

Offers high filtration efficiency and excellent filtrate purity

  • Allows usage with broad range of solutions, pH, and temperature
  • High flow and high retention capacity
  • Able to be sterilized by autoclaving with steam at 121°C for 20 minutes
  • Manual vent with luer lock to bleed air from upstream or serve as an injection or sample port

Specifications

Description Polycap HD 150 Capsule Filter, 0.45 μm, with FNPT inlet and outlet (5 pcs)
Pore Size 0.45 μm
For Use With (Application) Polycap HD 150
Housing Material Polypropylene
Inlet Style With Female NPT inlet/outlet
Max. Pressure 4.1 bar (60 psi)
Quantity 5/Pk.