missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polycap Disposable Capsules, 75 AS

Catalog No. 09927231
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Inlet Style:
Female National Pipe Thread
Stepped Barb
Outlet Style:
Female National Pipe Thread
Stepped Barb
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Inlet Style Outlet Style Sterility
09927231 0.45 μm Stepped Barb Stepped Barb Non-sterile
09927842 0.2 μm Female National Pipe Thread Female National Pipe Thread Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09927231 Supplier Cytiva Supplier No. 2707NS
Only null left

Polycap 75 AS filters for filtration of aqueous solutions

  • Sterile and nonpyrogenic
  • Integrity testable
  • With inlet vent
  • Filter Media: Low-extractable, hydrophilic nylon membrane and glass microfiber prefilter

  • Filling bell model (09-744-61) minimizes contamination when filtering into bottle or flask

Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.

Specifications

Autoclavable Autoclavable
Description Polycap AS 75 Capsule Filter, 0.45 μm, SB inlet and outlet (5 pcs)
Material (Membrane) Nylon
Pore Size 0.45 μm
Sterility Non-sterile
For Use With (Application) Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
Housing Material Polypropylene
Inlet Style Stepped Barb
Max. Pressure 4.1 bar (60 psi)
Outlet Style Stepped Barb