missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polycap™ Disposable Capsules - 36, 75, and 150 SPF

Provides serum prefiltration. Whatman™ Polycap™ Disposable Capsules permit filtration of tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspension. Includes inlet vent.

Supplier:  Cytiva 67053600

 View more versions of this product

Catalog No. 09-744-45


Only null left
Explore available promotions

Description

Description

  • Three-layer filter permits filtration of larger volumes
  • Integrity testable
  • With inlet vent
  • Filter Media: Three layers: fine glass microfiber/ultrafine glass microfiber/1.0μm polysulfone

Serum Prefilter Capsules. Retention (with air purge), <2mL

For serum prefiltration. Handle tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspensions.

Specifications

Specifications

Autoclavable
Glass Microfiber, Polyethersulfone
260 cm2
Polypropylene
Stepped Barb
Polycap SPF 36 Capsule Filter, 1.0 μm, with SB inlet and outlet (1 pc)
1 μm
For serum prefiltration. Handle tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspensions.
4.1 bar (60 psi)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions