missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Grade 17 Chr Cellulose Chromatography Paper

17 Chr cellulose chromatography roll is a 0.92 mm paper highly absorbent paper for preparative paper chromatography and electrophoresis.

$180.46 - $1854.00

Specifications

Description 17 Chr cellulose chromatography roll is a 0.92 mm paper highly absorbent paper for preparative paper chromatography and electrophoresis.
Flow Rate 190 mm/30 min.
Film Thickness 0.92 mm
For Use With (Application) Protein and Nucleic Acid Blotting, Preparative Paper Chromatography and Electrophoresis
Grade Grade 17 Chr
View More Specs

Products 3
Catalog Number Mfr. No. Length (Metric) Width (Metric) Format Price Quantity  
Catalog Number Mfr. No. Length (Metric) Width (Metric) Format Price Quantity  
09927702
View Documents
Cytiva
30178793
22 cm 2.5 cm Sheet
Pack of 100 Each for $200.00
Only null left
 
09927704
View Documents
Cytiva
3017621
30 cm 2.5 cm Roll
Each for $180.46
Only null left
 
09924555
View Documents
Cytiva
3017917
57 cm 46 cm Sheet
Pack of 100 Each for $1,854.00
Only null left
 
Description

Description

  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
  • Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
  • Also widely used in protein and nucleic acid blotting.
Specifications

Specifications

17 Chr cellulose chromatography roll is a 0.92 mm paper highly absorbent paper for preparative paper chromatography and electrophoresis.
0.92 mm
Grade 17 Chr
190 mm/30 min.
Protein and Nucleic Acid Blotting, Preparative Paper Chromatography and Electrophoresis
Cellulose
Videos
SDS
Documents

Documents