Learn More
Cytiva Whatman™ Cellulose Extraction Thimbles
Ensures optimal fit with its wide range of sizes suitable for use in thimble holders and automated extraction apparatus. Whatman™ Cellulose Extraction Thimbles are constructed of high-quality cellulose for excellent mechanical strength and retention.
Supplier: Cytiva 10350287
Description
Ensures optimal fit with its wide range of sizes suitable for use in thimble holders and automated extraction apparatus. Whatman™ Cellulose Extraction Thimbles are constructed of high-quality cellulose for excellent mechanical strength and retention.
- For all automated extraction apparatus
- High-quality cellulose material
- Dimensions matched exactly to thimble holders
Order Info
Extra allowance for wall thickness should be made when calculating external diameters
Chemical Identifiers
| 603 Standard Cellulose Extraction Thimbles made from high-quality cellulose available in a range of sizes. | |
| For the analysis of fats or pesticides in foods and soil materials as well as in many other procedures that involve a solid-liquid extraction | |
| 248°F | |
| Extraction Thimble |
| Cellulose | |
| For automated extraction apparatuses | |
| 130°C |
Specifications
| 603 Standard Cellulose Extraction Thimbles made from high-quality cellulose available in a range of sizes. | |
| 75 mm | |
| 2.5 mm | |
| 603 | |
| 9.8 in. | |
| 248°F | |
| Extraction Thimble |
| 2.9 in. | |
| Cellulose | |
| For the analysis of fats or pesticides in foods and soil materials as well as in many other procedures that involve a solid-liquid extraction | |
| For automated extraction apparatuses | |
| 250 mm | |
| 130°C |