Learn More
Cytiva Whatman™ Cellulose Extraction Thimbles
Ensures optimal fit with its wide range of sizes suitable for use in thimble holders and automated extraction apparatus. Whatman™ Cellulose Extraction Thimbles are constructed of high-quality cellulose for excellent mechanical strength and retention.
Supplier:  Cytiva 10350227
 View more versions of this product
Catalog No. 09924225
Get 1 Consumable Product When You Buy 1
View all promotions
Only null left
Description
Description
Ensures optimal fit with its wide range of sizes suitable for use in thimble holders and automated extraction apparatus. Whatman™ Cellulose Extraction Thimbles are constructed of high-quality cellulose for excellent mechanical strength and retention.
  • For all automated extraction apparatus
  • High-quality cellulose material
  • Dimensions matched exactly to thimble holders
Order Info
Extra allowance for wall thickness should be made when calculating external diameters
Specifications
Chemical Identifiers
603 Standard Cellulose Extraction Thimbles made from high-quality cellulose available in a range of sizes.
For the analysis of fats or pesticides in foods and soil materials as well as in many other procedures that involve a solid-liquid extraction
248°F
Extraction Thimble
Cellulose
For automated extraction apparatuses
130°C
Specifications
603 Standard Cellulose Extraction Thimbles made from high-quality cellulose available in a range of sizes.
28 mm
1.5 mm
603
3.9 in.
248°F
Extraction Thimble
1.10 in.
Cellulose
For the analysis of fats or pesticides in foods and soil materials as well as in many other procedures that involve a solid-liquid extraction
For automated extraction apparatuses
100 mm
130°C
Product Suggestions
Product Suggestions