Learn More
Cytiva Whatman™ AS 600 Series Multiple Vacuum Filtration Apparatus
Catalog No. p-4703914
Change view
Click to view available options
Volume (Metric):
500 mL
Includes:
Stainless Steel support screen filter, 6 place manifold, rapid closure clamp
Catalog No. Volume (Metric) Includes
09927413 500 mL Stainless Steel support screen filter, 6 place manifold, rapid closure clamp
1 options
Catalog No. 09927413 Supplier Cytiva Supplier No. 10444835
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AS610/3 Multiple Vacuum Filtration Apparatus, six-place vacuum filtration system, stainless steel 500 ml, 47/50 mm, support screen, with rapid closure clamp
AS 600 Series
The apparatus can be autoclaved and sterilized by dry heat at up to 180°C. Suitable only for vacuum operation. If flushing tubes are used, do not exceed 1.3 bar (300 mbar over-pressure).
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Includes Stainless Steel support screen filter, 6 place manifold, rapid closure clamp
Series AS610/3 Series
Type Vacuum Filtration System
Volume (Metric) 500 mL