missing translation for 'onlineSavingsMsg'
Learn More

Abcam Western Blot Stripping Buffer, Liquid, 500 mL

Numéro de catalogue. 502655343
Encompass
Modifier l'affichage
Click to view available options
Quantité:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
502655343 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 502655343 Fournisseur Abcam Code fournisseur AB282569500ML

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Uniformly removes antibodies from Western blots for reprobing.

  • Form: Liquid
  • Storage: Store at Ambient
  • Shipping: Ambient - Cannot Ship with Ice
  • pH: 2.9 to 3.1
  • Constituents: 88% Water, 10% Sodium lauryl sulfate
  • Application: Uniformly removes antibodies from Western blots for reprobing

The product is subject to the following: Abcam Restricted Use Statement

Spécifications

Shipping condition (Condition d'expédition) Ambient
Format Bottle
Tampon 10% sodium lauryl sulfate, pH 2.9 to 3.1
Contenu et stockage Store at ambient temperature.
Compatibilité des membranes Nitrocellulose, PVDF
Type de produit Wash Solution
Forme Liquid
Formule pH 2.9 to 3.1, 88% Water, 10% Sodium lauryl sulfate
Quantité 500 mL
À utiliser avec (application) Western Blot