Learn More
Wells Lamont™ Scepter™ Sterile Antimicrobial M321 Cut-Resistant Gloves
Lightweight and sterile gloves protect hands from prolonged exposure to latex and have a high level of tactile sensation. Scepter™ Sterile Antimicrobial M321 Cut-Resistant Gloves can be used as a liner or stand alone glove.
Supplier:  Wells Lamont™ M321S
 View more versions of this product
Catalog No. 17200333
Only null left
Explore available promotions
Description
Description
  • Stainless steel and polyester construction
  • Launderable 15 times without shrinking
  • Moderate tactile sensitivity
  • Antimicrobial
  • Sterile and ambidextrous
  • Applications: Medical, organ tissue processing and pharmaceutical
  • ANSI Cut Level 4
  • Made in the USA
Specifications
Specifications
ASTM F1790-97, ASTM F2992-15
Uncoated
ANSI A4
Stainless Steel Fiber, Polyester
Small
Medical, organ tissue processing, pharmaceutical
Green
Sceptor™ M321
Category III
White
Ambidextrous
Cut-resistant Gloves
Sterile
Green
M321S
Product Suggestions
Product Suggestions