Learn More
Wells Lamont™ Cut-Resistant FR Sleeves
Composite Cut-Resistant FR Sleeves offer a high level of cut protection, while also providing outstanding comfort and fit. Made of a combination of high performance materials and flame resistant fibers, which will allow it to be used where incidental protection against flame and sparks are required.
Supplier:  Wells Lamont™ SKC18
 View more versions of this product
Catalog No. 19148972
Only null left
Explore available promotions
Description
Description
  • Unique knit sleeve made from a patented combination of high performance materials and flame resistant fibers
  • Knitted using strong para-aramid yarns
  • Lightweight construction will increase employee acceptance and reduce the risk of injury
  • Blousy fit allows superior comfort and breathability
  • Elastic on both ends allows sleeve to stay in place
  • Available in several lengths, with or without a thumb hole, and also a wide top design for larger biceps
  • ANSI Cut Level A3
  • ANSI Flame Rating 4
  • Sold by the dozen pieces
  • Made in the USA
Specifications
Specifications
Lightweight high performance materials and flame resistant fiber
White, Yellow, Black
Elastic Ends
Knit
Used to increase employee acceptance and reduce the risk of injury
45.72 cm
Cut-resistant Sleeve
Standard Sleeve
ANSI Level 3
Laundered using a mild soap and detergent, use a warm water wash with a temperature not to exceed 160°F, wash time should not be longer than 15 to 20 min, use a rinse cycle for the product in cold water, use a high extraction speed, tumb
18 in.
Product Suggestions
Product Suggestions