missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Weighing Pan

Catalog No. 14558726
Change view
Click to view available options
Capacity (Metric):
100 to 250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14558726 100 to 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14558726 Supplier Sartorius Supplier No. YWP10

Titanium Weighing Pan for Cubis™ I High-Capacity Microbalances.

  • Weighing Modules: MSA 66S, MSA 66P and MSA 116P

Specifications

Autoclavable Not Autoclavable
Bottom Shape Round
Capacity (Metric) 100 to 250 mL
Color Silver
Disposable Reusable
Material Titanium
Shape Round
Sterility Non-sterile
Type Weighing Dish
Size 15.36 cm3
Diameter (Metric) 90 mm
Show More Show Less