Learn More
Invitrogen™ WedgeWell™ Empty Gel Cassettes with Combs, midi
Description
These WedgeWell empty gel cassettes are pre-assembled, single-use, plastic cassettes with combs that allow you to pour your own midi-size gels. The gels can be run in the SureLock Tandem Midi Gel Tank (STM1001) or XCell4 SureLock Midi-Cell (WR0100). The cassettes are sealed on three sides to prevent leaking and do not require the use of a special casting stand. Gels poured in these cassettes are 1.0-mm thick. SureCast reagents can used to make pouring your own high performance polyacrylamide midi gels both easy and reproducible.
These WedgeWell empty gel cassettes are pre-assembled, single-use, plastic cassettes with combs that allow you to pour your own midi-size gels. The gels can be run in the SureLock Tandem Midi Gel Tank (STM1001) or XCell4 SureLock Midi-Cell (WR0100). The cassettes are sealed on three sides to prevent leaking and do not require the use of a special casting stand. Gels poured in these cassettes are 1.0-mm thick. SureCast reagents can used to make pouring your own high performance polyacrylamide midi gels both easy and reproducible.
Product features
- The cassettes are sealed on three sides to prevent leaking and do not require the use of a special casting stand
- Gels poured in these cassettes are 1.0-mm thick
- SureCast reagents can be used to pour high-performance polyacrylamide midi gels
Handcast gel reagents
SureCast protein gels are a reliable and high-performance solution for your handcast gel electrophoresis needs. SureCast resolving and stacking buffers are based on the traditional Laemmli protein chemistry, allowing the use of Laemmli sample and running buffers. Handcast SureCast gels are a reliable, high-performance, low-cost option for gel electrophoresis.
Convenient to use
The midi WedgeWell empty gel cassettes have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, which makes sample loading easier.
Specifications
Specifications
| Content And Storage | 10 cassettes (with 12+2 well combs) per pack |
| For Use With (Equipment) | XCell4 SureLock Midi-Cell |
| Gel Size | Midi |
| Type | Empty Gel Cassette |
| No. of Wells | 12 + 2-well |
| Quantity | 1 pack |
| Running Dimension | Vertical |
| Shipping Condition | Room Temperature |
| Thickness (Metric) | 1.0 mm |
| Length (Metric) Gel | 8 cm |
| Show More |
Frequently Asked Questions (FAQs)
For the 12+2 well large gels, the maximum sample loading volume is 100 µL. For the 12+2 well small gels, the maximum sample loading volume is 90 µL. For 20 well gels, the maximum sample loading volume is 60 µL and for the 26 well gels, the maximum sample loading volume is 40 µL.
We do offer disposable, empty plastic cassettes and combs to provide the option of pouring your own Mini or Midi gels for use with the XCell SureLock Mini-Cell, Mini Gel Tank, or XCell4 SureLock Midi-Cell, respectively. All cassettes are sealed on three sides to prevent leaking. Since the cassettes are pre-sealed with the slot pre-taped, a bulky casting stand is not necessary. The empty cassettes and combs can be found here (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/power-supplies-protein-electrophoresis.html#staining).
- The Empty Mini Gel Cassettes are available in two thicknesses (1.0 mm and 1.5 mm) and can be used with Invitrogen Combs that are available in two thicknesses (1.0 mm and 1.5 mm), and in 7 types (1 well, 2 well, 5 well, 10 well, 12 well, 15 well, and 2D well).
- The Bolt Empty Mini Gel Cassettes are available in 1.0 mm thickness and can be used with Bolt 10- or 12-well combs.
- The Empty Midi Gel Cassettes are available in 1.0 mm thickness and can be used with three Invitrogen Comb types (12 + 2 wells, 20 wells, and 26 wells).
For Research Use Only. Not for use in diagnostic procedures.