Learn More
Thermo Scientific™ WebSeal™ 96-Well Plate Kits with Individual Closures
Save time and minimize the risk of vial contamination prior to use with 96-Well Plate Kits packaged with vials sealed with individual closures.
$165.60 - $179.44
Specifications
Format Standard
Array 8 x 12
Product Line Webseal
Color Clear
Product Type Glass Inserted Plate Kit with Individual Closures
View More Specs
Products 3
Catalog Number Mfr. No. Total Volume (Metric) Well Lid Price Quantity & Availability  
Catalog Number Mfr. No. Total Volume (Metric) Well Lid Price Quantity & Availability  
60105419
View Documents Promotion Details
Thermo Scientific™
60180K201
400 μL Silicone/PTFE, Pre-slit 45° Septa
Pack for $168.49
Only null left
60105420
View Documents Promotion Details
Thermo Scientific™
60180K202
1200 μL Silicone/PTFE, 45° Septa
Pack for $165.60
Only null left
60105421
View Documents Promotion Details
Thermo Scientific™
60180K203
1200 μL Silicone/PTFE, Pre-slit 45° Septa
Pack for $179.44
Only null left
Description
Description
Save time and minimize the risk of vial contamination prior to use with Thermo Scientific™ WebSeal™ 96-Well Plate Kits with Individual Closures. Compatible with all 96-well-configured autosamplers, the ready-to-use kits consist of a comprehensive choice of 96-well microplates and glass vials with pre-attached caps and septa.
Specifications
Specifications
Standard
Webseal
Glass Inserted Plate Kit with Individual Closures
1
8 x 12
Clear
96
1/Pk.