missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR44 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
This gene encodes a protein that interacts with the small GTPase rab11. A similar protein in rat binds the GTP-containing active form of rab11. This protein may play a role in endosome recycling. Alternate splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | WDR44 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | WDR44 |
| Gene Accession No. | Q5JSH3 |
| Gene Alias | WD repeat domain 44, RAB11BP, RPH11 |
| Gene Symbols | WDR44 |
| Host Species | Rabbit |
| Show More |