missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WDR1 Recombinant Rabbit Monoclonal Antibody (23GB4720)
GREENER_CHOICE

Catalog No. MA553796
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553796 100 μL
MA553797 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553796 Supplier Invitrogen™ Supplier No. MA553796
Only null left

Rabbit Recombinant Monoclonal Antibody

WDR1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

This gene encodes a protein containing 9 WD repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, mostly including a trp-asp at the C-terminal end. WD domains are involved in protein-protein interactions. The encoded protein may help induce the disassembly of actin filaments. Two transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen WDR1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4720
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene WDR1
Gene Accession No. O75083, O88342, Q5RKI0
Gene Alias Actin-interacting protein 1; Aip1; D5Wsu185e; epididymis secretory protein Li 52; HEL-S-52; NORI-1; rede; WD repeat domain 1; WD repeat protein 1; WD repeat-containing protein 1; WD40 repeat protein 1; Wdr1
Gene Symbols WDR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human WDR1 (321-350AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22388, 360950, 9948
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less