Learn More
Watson-Marlow Single-Segment Manifold Tubing for Peristaltic Pump Models 205S/CA and 205U/CA: Marprene
For use with Peristaltic Pump Models 205S/CA and 205U/CA.
$215.73 - $370.78
Specifications
Material Marprene
Thickness (English) 0.03 in.
Thickness (Metric) 0.8 mm
For Use With 205S/CA and 205U/CA peristaltic pump
Min. Operating Temperature (English) 41°F
View More Specs
Products 13
Catalog Number Mfr. No. Tubing Size Flow Rate Color Price Quantity & Availability  
Catalog Number Mfr. No. Tubing Size Flow Rate Color Price Quantity & Availability  
1428389
View Documents Promotion Details
Watson-Marlow
9780254 000
2.54mm 0.010 to 18mL/min. Purple/Orange
Pack of 6 Each for $215.73
Only null left
1428374
View Documents Promotion Details
Watson-Marlow
9780025 000
0.25mm 0.001 to 0.265mL/min. Orange/Blue
Pack of 6 Each for $215.73
Only null left
1428375
View Documents Promotion Details
Watson-Marlow
9780038 000
0.38mm 0.001 to 0.439mL/min. Orange/Green
Pack of 6 Each for $215.73
Only null left
1428379
View Documents Promotion Details
Watson-Marlow
9780088 000
0.88mm 0.014 to 2.520mL/min. Orange/Orange
Pack of 6 Each for $257.58
Only null left
1428381
View Documents Promotion Details
Watson-Marlow
9780114 000
1.14mm 0.023 to 4.1mL/min. Red/Red
Pack of 6 Each for $215.73
Only null left
1428383
View Documents Promotion Details
Watson-Marlow
9780142 000
1.42mm 0.035 to 6.25mL/min. Yellow
Pack of 6 Each for $215.73
Only null left
1428380
View Documents Promotion Details
Watson-Marlow
978.0102.000
1.02mm 0.019 to 3.033mL/min. White
Pack of 6 Each for $227.38
Only null left
1428376
View Documents Promotion Details
Watson-Marlow
9780050 000
0.5mm 0.004 to 0.720mL/min. Orange/Yellow
Pack of 6 Each for $215.73
Only null left
1428384
View Documents Promotion Details
Watson-Marlow
9780152 000
1.52mm 0.04 to 7.15mL/min. Yellow/Blue
Pack of 6 Each for $215.73
Only null left
1428385
View Documents Promotion Details
Watson-Marlow
9780165 000
1.65mm 0.046 to 8.26mL/min. Blue/Blue
Pack of 6 Each for $215.73
Only null left
1428390
View Documents Promotion Details
Watson-Marlow
9780279 000
2.79mm 0.117 to 21mL/min. Purple/White
Pack of 6 Each for $370.78
Only null left
1428387
View Documents Promotion Details
Watson-Marlow
9780205 000
2.05mm 0.69 to 12.4mL/min. Purple/Purple
Pack of 6 Each for $370.78
Only null left
1428386
View Documents Promotion Details
Watson-Marlow
9780185 000
1.85mm 0.057 to 10.3mL/min. Green/Green
Pack of 6 Each for $215.73
Only null left
Description
Description
For use with Peristaltic Pump Models 205S/CA and 205U/CA.
Order Info
Comes in packs of six single segments.
Specifications
Specifications
Marprene
0.8 mm
41°F
176°F
0.03 in.
205S/CA and 205U/CA peristaltic pump
5°C
80°C