Learn More
Watson-Marlow Manifold Tubing - PVC
Double pump segment manifold tubing — use one, transfer to the other for double life
Supplier: Watson-Marlow 981 0152 000
Description
Double pump segment manifold tubing — use one, transfer to the other for double life
- For use with 400DM2 and 400DM3 pumpheads
- Can be used with any multichannel cartridges
- Adjacent cartridges may contain tubing of a different material or size
Chemical Identifiers
| PVC | |
| 0.8 mm | |
| Pump Tubing |
| 0.03 in. | |
| 323S/MC peristaltic pump |
Specifications
| 1/20 in. | |
| PVC | |
| 0.8 mm | |
| 1.52mm | |
| Yellow/Blue |
| 1.52 mm | |
| 0.03 in. | |
| 323S/MC peristaltic pump | |
| 0.27 to 19 (four roller pump), 0.2 to 14 (eight roller pump)mL/min. | |
| Pump Tubing |