Learn More
Watson-Marlow Manifold Tubing - PVC
Double pump segment manifold tubing — use one, transfer to the other for double life
Supplier: Watson-Marlow 981 0063 000
Description
Double pump segment manifold tubing — use one, transfer to the other for double life
- For use with 400DM2 and 400DM3 pumpheads
- Can be used with any multichannel cartridges
- Adjacent cartridges may contain tubing of a different material or size
Chemical Identifiers
| PVC | |
| 0.8 mm | |
| Pump Tubing |
| 0.03 in. | |
| 323S/MC peristaltic pump |
Specifications
| 1/50 in. | |
| PVC | |
| 0.8 mm | |
| 0.63mm | |
| Orange/White |
| 0.63 mm | |
| 0.03 in. | |
| 323S/MC peristaltic pump | |
| 0.04 to 3.1 (four roller pump), 0.04 to 2.6 (eight roller pump)mL/min. | |
| Pump Tubing |