missing translation for 'onlineSavingsMsg'
Learn More

Oakton™ Waterproof PD 450 Portable pH/DO Meter

Obtain pH and dissolved oxygen readings from one meter.

Supplier:  Oakton™ 3563230

 View more versions of this product

Catalog No. 15500144


Only null left

Description

Description

The PD 450 meter uses optical measurement technology offering fast response times and long-term stability with minimal maintenance for dissolved oxygen measurements.

  • Optical DO sensor eliminates need for tiresome membranes and solutions
  • Durable waterproof and dustproof design with IP67 rating makes this meter ideal for plant or field testing
  • Large, backlit LCD to view pH, mV, % saturation or mg/L (ppm) simultaneously with temperature
  • Automatic temperature compensation, barometric pressure compensation and salinity correction ensures DO measurement accuracy
  • Memory for up to 500 data sets with USB or RS-232 output to PC
  • Additional features include: ready (stability) indicator, smart averaging, hold function, calibration alarm and battery-life indicator
  • Perform data logging with manual or timed collection of up to 500 data sets with real-time clock time/date stamp
  • Built-in stand allows for easy benchtop use

Specifications:

  • pH Range: -2.00 to 16.00
  • pH Resolution: 0.01
  • pH Accuracy: ±0.01
  • mV Range: ±2000
  • mV Resolution: 0.01
  • mV Accuracy: ±0.5% full scale
  • DO Range: 0 to 20ppm; 0 to 200%
  • DO Resolution:0.1 ppm; 0.1%
  • DO Accuracy: ±2%
  • Temperature Range: -10° to +110°C (17° to 230°F)
  • Temperature Resolution: 0.1
  • Temperature Accuracy: ±0.5
  • Temperature Compensation: Automatic or manual
  • Interface: USB or RS-232
  • Calibration: Up to six points
  • Buffer Recognition: USA, NIST, DIN, custom
  • Power: two AA batteries or optional AC adapter
  • Battery Life: 500 hours
  • Dimensions (L x W x H): 20 x 8 x 6.3cm (7.9 x 3.12 x 2.5 in.)

Includes:

  • Cat. No. 15-500-144: PD 450 meter, pH probe, RDO probe, sensor cap and two AA batteries
  • Cat. No. 15-500-145: PD 450 meter, pH probe, RDO probe, sensor cap, two AA batteries and NIST-traceable calibration certificate provided by InnoCal
  • Cat. No. 15-500-146: PD 450 meter, pH probe, RDO probe, sensor cap, calibration solutions, two AA batteries, and hard carrying case
  • Cat. No. 15-500-147: PD 450 meter, pH probe, RDO probe, sensor cap, calibration solutions, two AA batteries, hard carrying case and NIST™-traceable calibration certificate provided by InnoCal

Order Info

Order optional data cable for USB or RS-232 connectivity to a PC separately.

Certifications

CE marked, TUV 3-1, FCC Class A

Warranty

Three years

Specifications

Chemical Identifiers

±0.5% full scale
±2%
Up to 6
500 Sets
IP67
-2 to 16
-10°C to +110°C
0 to 20ppm; 0 to 200%
0.1 ppm; 0.1%
17°F to 230°F
3 Years
20 cm
8 cm
6.3 cm
±0.01
2 AA
CE marked, TUV 3-1, FCC Class A
Backlit LCD
±2000
0.01
Ideal for plant or field testing.
0.01
±0.5
0.1
7.9 in.
3.12 in.
2.5 in.

Specifications

±0.5% full scale
±2%
Up to 6
500 Sets
IP67
-2 to 16
-10°C to +110°C
PD 450 meter, pH probe, RDO probe, sensor cap and two AA batteries
0 to 20ppm; 0 to 200%
0.1 ppm; 0.1%
17°F to 230°F
3 Years
20 cm
8 cm
6.3 cm
±0.01
2 AA
CE marked, TUV 3-1, FCC Class A
Backlit LCD
±2000
0.01
Ideal for plant or field testing.
Meter with Probes
0.01
±0.5
0.1
7.9 in.
3.12 in.
2.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions