missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Water, WFI Quality, Sterile Purified Water, Cell Culture Tested, OmniPur™, Calbiochem™,

Catalog No. p-7082968
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4865050500 500 mL
4865051000 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4865050500 Supplier MilliporeSigma™ Supplier No. 4.86505.0500
Only null left

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Packaging Plastic Bottle
Quantity 500 mL
Physical Form Liquid