missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, USP/EP Purified, Ricca Chemical
Description
Water, USP/EP Purified
Specifications
Specifications
| pH | 5.0 to 7.0 |
| Color | Clear, Colorless |
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Conductivity | max. 1 μS/cm |
| Endotoxin | max. 0.25 EU/mL at time of manufacture |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| IUPAC Name | oxidane |
| Molecular Weight (g/mol) | 18.015 |
| Percent Purity | 100% |
| Show More |
Safety and Handling
| ShelfLife | 12 months |