missing translation for 'onlineSavingsMsg'
Learn More

Water, USP/EP Purified, Ricca Chemical

Catalog No. 91905HP
Change view
Click to view available options
Quantity:
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Hard Pack
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
91905HP 20 L Hard Pack
91901G 4 L Amber Glass Bottle
91901 4 L Natural Poly Bottle
919025 10 L Cubitainer
91905 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 91905HP Supplier Ricca Chemical Company Supplier No. 91905HP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Water, USP/EP Purified

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 962
ChEBI CHEBI:15377
SMILES O

Specifications

pH 6
Color Colorless
Melting Point 0.0°C
CAS Min % 100
Boiling Point 100.0°C
CAS Max % 100
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Miscible in water
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
Formula Weight 18.02
Percent Purity 100%
Grade EP/USP
Packaging Hard Pack
Density 1
Quantity 20 L
Physical Form Liquid
Chemical Name or Material Water, USP/EP Purified
Show More Show Less
ShelfLife 12 months
Recommended Storage Ambient