missing translation for 'onlineSavingsMsg'
Learn More

Water, USP/EP Purified, Reagents

Catalog No. p-200020873
Change view
Click to view available options
Quantity:
20 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336897 200 L
01336896 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336897 Supplier Labchem Inc Supplier No. C2346100200E
Only null left

Water, USP/EP Purified, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Dihydrogen Oxide
IUPAC Name water
SMILES O

Specifications

pH Neutral
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Percent Purity 100%
Grade USP
Packaging Poly Drum
Sterility Non-sterile
Quantity 200 L
Specific Gravity 1
Physical Form Liquid
Chemical Name or Material Water, USP/EP Purified
Show More Show Less