missing translation for 'onlineSavingsMsg'
Learn More

Water, USP/EP Purified, Reagents

Catalog No. 01336896
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336896 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336896 Supplier Labchem Inc Supplier No. C234610020F
Only null left

Water, USP/EP Purified, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Dihydrogen Oxide
IUPAC Name water
SMILES O

Specifications

pH Neutral
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Percent Purity 100%
Grade USP
Packaging Cubitainer
Sterility Non-sterile
Quantity 20 L
Specific Gravity 1
Physical Form Liquid
Chemical Name or Material Water, USP/EP Purified
Show More Show Less