missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, USP/EP/JP Purified, Ricca Chemical
USP/EP Purified Water
$274.75 - $274.75
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
| PubChem CID | 962 |
| ChEBI | CHEBI:15377 |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R918900020E
|
Ricca Chemical Company
R918900020E1 |
20 L | Hard Pack |
N/A
|
|
||||
Chemical Identifiers
| 7732-18-5 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| 962 | |
| O |
| H2O | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
| CHEBI:15377 |
Specifications
| 6 | |
| Colorless | |
| 100 | |
| 100 | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| oxidane | |
| 962 | |
| 18.02 | |
| EP/JP/USP | |
| 1 | |
| Liquid |
| 7732-18-5 | |
| 0°C | |
| 100°C | |
| H2O | |
| Miscible in water | |
| O | |
| 18.015 | |
| CHEBI:15377 | |
| 100% | |
| Hard Pack | |
| 20 L | |
| Water |
Safety and Handling
ShelfLife : 12 months
Recommended Storage : Ambient