missing translation for 'onlineSavingsMsg'
Learn More

Water Ultrapur, MilliporeSigma™

Catalog No. p-7162463
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1012620500 500 mL
361044623 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1012620500 Supplier Technidata Supplier No. 1.01262.0500
Only null left

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Deionized water
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C (1013 hPa)
Packaging Plastic Bottle
Vapor Pressure 23 hPa (20°C)
Density 1.00 g/cm3 (at 20°C)
Quantity 500 mL
Recommended Storage Store at 5 to 30°C