missing translation for 'onlineSavingsMsg'
Learn More

Water, Ultra Trace Elemental Analysis Grade, Fisher Chemical™

Packaged in specially precleaned LDPE bottles to protect product integrity

Supplier:  Seastar Chemicals Inc S020901TFIL07

 View more versions of this product

Catalog No. W92


Only null left
Explore available promotions

Description

Description

  • Evaluated for 65 metal impurities in ppt range
  • Also specified for ≤1ppb for chloride, phosphate and sulfate anion impurities
  • Packaged in precleaned LDPE bottles to maintain product integrity
Specifications

Specifications

Water
Colorless
0°C
H2O
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
Ultra Trace Elemental Analysis
17.5mmHg at 20°C
Certificate of Lot Analysis included with each bottle.
Liquid
7
7732-18-5
100°C
11332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
LDPE Bottle
1.000g/cm³
2 L

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
11332
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions